Would you like to log in again?
Cell Meter™ Fluorimetric Phagocytosis Assay Kit *Red Fluorescence*, AAT Bioquest
Supplier: AAT Bioquest
21225
76484-426EA 361.89 USD InStock
76484-426
Cell Meter™ Fluorimetric Phagocytosis Assay Kit *Red Fluorescence*, AAT Bioquest
Assays Cellular Assays
Phagocytosis is defined as the cellular uptake of particles within a plasma-membrane envelope by a cell.
  • Ready to use in suspension
  • No trypan blue quenching required
  • Simultaneous detection of live cells/phagocytosis
  • Suitable for all major fluorescence platforms
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty