Would you like to log in again?
OxiVision™ Green Hydrogen Peroxide Sensor (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21505
76483-414EA 118.54 USD InStock
76483-414
OxiVision™ Green Hydrogen Peroxide Sensor (1 mg), AAT Bioquest
Stains Microbiology Stains
Despite the importance of H₂O₂ to human health and disease, the molecular mechanisms of its production, accumulation, trafficking, and function are insufficiently understood due to the lack of sensitive and specific H₂O₂ sensors that can be used in live cells.
  • Suitable for live cells
  • Extremely selective for Hydrogen peroxide
  • Bright signal
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty