Would you like to log in again?
Calcein UltraBlue™ Sodium Salt (1 mg), AAT Bioquest
Supplier: AAT Bioquest
21909
76483-460EA 240.68 USD InStock
76483-460
Calcein UltraBlue™ Sodium Salt (1 mg), AAT Bioquest
Stains Microbiology Stains
Calcein UltraBlue™ has similar spectral properties to those of Calcein Blue.
  • Low cytotoxicity
  • Improved photostability
  • Brighter fluorescence
  • Suitable for multiplexing
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty