Would you like to log in again?
ReadiUse™ 10KD Spin Filter (5), AAT Bioquest
Supplier: AAT Bioquest
60502
76485-620EA 89.37 USD InStock
76485-620
ReadiUse™ 10KD Spin Filter (5), AAT Bioquest
Filters Centrifugal Filters
ReadiUse™ 10 Kd Spin Filter is a disposable ultrafiltration device for the concentration of biological samples such as antibodies.
  • Compatible with both swing bucket or fixed angle instruments
  • Designed to prevent membrane fouling
  • Low internal surface area
  • High material recovery from dilute solutions
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty