Would you like to log in again?
Glass Beaker Set, Borosilicate, United Scientific Supplies
Supplier: United Scientific Supplies
BGSET5
76531-422EA 28.54 USD InStock
76531-422
Glass Beaker Set, Borosilicate, United Scientific Supplies
Beakers
Set of five borosilicate glass beakers: one each of 50 ml, 100 ml, 250 ml, 600 ml and 1000 ml capacities.
  • Feature pour spouts, double-graduated metric scale and durable marking area
This starter set of beakers comes with all the most commonly used sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty