Would you like to log in again?
Pipettes, Serological, Accuracy as per Class B, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PS7080-C PS7080-A PS7080-J PS7080-F PS7080-I PS7080-B PS7080-D PS7080-H
76532-124PK 27.29 USD InStock
76532-124 76532-122 76532-116 76532-128 76532-114 76532-126 76532-118 76532-120
Pipettes, Serological, Accuracy as per Class B, United Scientific Supplies
Pipettes Serological Pipettes
Type 3 serological pipettes with color-coding.
  • Printed with durable amber markings at intervals
  • Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with USP, Class B standards. Calibrated to deliver.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty