Would you like to log in again?
Graduated Cylinders, Class B, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
CY3022-1000 CY3022-25 CY3022-2000 CY3022-50 CY3022-500 CY3022-250
76531-622PK 176.67 USD InStock
76531-622 76531-624 76531-620 76531-618 76531-626 76531-628
Graduated Cylinders, Class B, Borosilicate Glass, United Scientific Supplies
Cylinders Measuring Cylinders
Fine quality borosilicate glass cylinders .
  • Features double metric scale
  • Stable hexagon base
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases. All cylinders come with bumper guards.
Certifications: Cylinders comply with ASTM E1272, Class B standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty