Would you like to log in again?
Acrylic Block, United Scientific
Supplier: United Scientific Supplies
RGF114-A
470332-634EA 2.5 USD InStock
470332-634
Acrylic Block, United Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
Illuminate the path of light through an acrylic block.
  • Acrylic
  • Two largest sides are frosted
When light passes though clear acrylic you can not see the exact path it travels. When a side is frosted it reflects the light so you can easily see the path that light travels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty