Would you like to log in again?
Digital Stopwatch
Supplier: United Scientific Supplies
STOPWD-DIG
470332-648EA 3.05 USD InStock
470332-648
Digital Stopwatch
Timers Digital Stopwatches
Digital stopwatch with lanyard.
  • Split and cumulative time modes
  • 1/100th second increments
  • Calendar/date display
  • Alarm
Includes a LR1130 coin battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty