Would you like to log in again?
Electrostatics Kit, United Scientific
Supplier: United Scientific Supplies
ELSKIT
470332-764EA 25.5 USD InStock
470332-764
Electrostatics Kit, United Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Convenient collection designed for investigations around charges created through friction.
  • Explore static charges
  • Create positive and negative charges
The Electrostatics Kit is a collection of materials designed for students to investigate the creation of charges through friction between various rods and fabrics.
Ordering information: Kit includes 1 hollow glass friction rod, 1 glass friction rod, half ground, 1 ebonite rod, 1 spool of string (10'), 3 balloons, 1 silk pad (approx. 6" square), 1 faux fur pad (approx. 6" square), and 1 flannel pad (approx. 6" square).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty