Would you like to log in again?
MacConkey Agar, MP Biomedicals
Supplier: MP Biomedicals
091006017
76461-576EA 134.87 USD InStock
76461-576
MacConkey Agar, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
A preparation for the isolation of enteric bacilli based on the fermentation of lactose.
Ready for CE IVD labeling of clinical applications.
Caution: Research use only (RUO).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty