Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc stearate

Supplier: Thermo Scientific Chemicals
Synonyms: Stearic acid zinc saltZinc(II) stearate

AC61151-5000
TS61151-5000EA 27.15 USD InStock
TS61151-5000
Zinc stearate
Zinc stearate


Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Melting Pt: 118…128 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/73/2302573.jpg
Order Now
Specification Test Results
Appearance White Powder
Assay 12.5 to 14.0 % (ZnO)
Identification Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty