Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicic acid hydrate, powder

Supplier: Thermo Scientific Chemicals

AC61123-5000
TS61123-5000EA 372 USD InStock
TS61123-5000
Silicic acid hydrate, powder
Silicic acid hydrate
Appearance: White Powder


Formula: SiO₂·xH₂O
MW: 60.09 g/mol (anhy)
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00054122
CAS Number: 1343-98-2
REACH: 01-2119379499-16
Order Now
Specification Test Results
Appearance White Powder
Identification Passes test
Non-volatile matter ≤0.40 % (in HF)
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.020 %
Iron (Fe) ≤0.01 %
Heavy metals (as Pb) ≤0.010 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty