Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (III) citrate 16.5 - 18.5% (Fe)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ammonium iron (III) citrateFerric Ammonium CitrateAmmonium ferric citrate

AC61221-5000
TS61221-5000EA 62.13 USD InStock
TS61221-5000
Ammonium iron (III) citrate 16.5 - 18.5% (Fe)
Ammonium iron (III) citrate


Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Order Now
Specification Test Results
Color Brown to Brown-Yellow
Physical Form Powder
Percent Purity 16.5 to 18.5% (Fe)
Identification Pass Test
Infrared Spectrum Authentic

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty