Would you like to log in again?
Media/Storage Bottles, GL 45, Square, Clear Plastic (PC)
Supplier: United Scientific Supplies
BMPC250 BMPC150 BMPC1000 BMPC500
470320-966PK 331.5 USD InStock
470320-966 470320-960 470320-962 470320-964
Media/Storage Bottles, GL 45, Square, Clear Plastic (PC)
Bottles Plastic Bottles
Clear polycarbonate bottles for storage.
  • Square space-saving design
  • Supplied with polypropylene cap
  • Sterilized using gamma radiation
  • Individually bagged
  • Autoclavable
Clear, square disposable polycarbonate bottles are space-saving and convenient to handle. Useful for mixing, sampling, and storage applications. Graduations and large marking spot are screened in easy-to-read white enamel. Includes a blue, linerless GL 45 polypropylene plug-seal autoclavable cap. Neck int.Ø is 37 mm.
Polycarbonate bottles can be used in environments ranging from −80...+130 °C. Bottles can be autoclaved once. Repeated autoclaving is not recommended.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty