Would you like to log in again?
AccuThrive Dermablade Biopsy Blade, Stainless Steel, Sterile, AccuTec Blades
Supplier: AccuTec Blades
AVSC-1130-0000
76538-996BX 150.19 USD InStock
76538-996
AccuThrive Dermablade Biopsy Blade, Stainless Steel, Sterile, AccuTec Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Shave biopsy blade.
  • Handles allow for controlled precise excisions
AccuThrive's MicroCoat blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp effective edge over multiple uses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty