Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutyltin 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TetrabutylstannaneTetra-n-butyltin

AC13798-0050
TS13798-0050EA 21 USD InStock
TS13798-0050
Tetrabutyltin 96%
Tetrabutyltin


Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145 °C (10 mmHg)
Melting Pt: –97 °C
Density: 1.057 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 2788
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
GC ≥95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty