Would you like to log in again?
Impervious Comfort Gowns, Halyard
Supplier: Halyard
69602 69600 69601 69603
76540-402CS 330.12 USD InStock
76540-402 76540-400 76540-406 76540-404
Impervious Comfort Gowns, Halyard
Disposable Garments
Offer protection from blood and body fluids and help maintain the health and confidence of personnel.
  • Perforated open-back design
  • Knit cuffs
  • Film laminate
These non sterile gowns feature film laminate, poly-coated spunbond. The garments combine effective fluid resistance with wearer comfort. These gowns are ideal for patient contact, fluid isolation, decontamination, or general cleanup tasks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty