Would you like to log in again?
Corning® Laboratory Bottles, Low Actinic, Ace Glass
Supplier: Ace Glass
Corning®
4049-21 4049-21 4049-12 4049-18 4049-04 4049-10 4049-25 4049-34 4049-04 4049-06 4049-25 4049-10 4049-08 4049-18 4049-08 4049-34 4049-12 4049-06
76529-614CS 366.83 USD InStock
76529-614 76529-616 76529-606 76529-618 76638-908 76529-608 76529-620 76529-610 76529-622 76529-612 76638-896 76638-910 76638-898 76638-894 76638-904 76638-906 76638-900 76638-902
Corning® Laboratory Bottles, Low Actinic, Ace Glass
Bottles Glass Bottles
Media storage bottles with low actinic stain fired into the exterior surface.
  • Heavy-duty bottle of low actinic glass
  • Excellent for storing/transporting light senstive materials
  • Autoclavable
Protect materials sensitive to light in the 200 to 500 nanometer range. Glass bead indicates full capacity line. Permanent white enamel graduations and marking spots included.
Delivery information: Includes a linerless, one-piece, autoclavable (120 °C), orange polypropylene, plug-seal cap with drip-free pouring rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty