Would you like to log in again?
Tubing Connector; with Ace-Threds, Ace Glass
Supplier: Ace Glass
5858-214
76529-630EA 66.92 USD InStock
76529-630
Tubing Connector; with Ace-Threds, Ace Glass
Fittings and Connectors
Glass filled PTFE tubing connector for use with Ace-Thred connections.
  • Glass filled PTFE for mechanical strength and chemical inertness
  • Threaded connection with O-Ring seal
  • Safety and flexibility improvement over glass hose connections
  • Stronger than PFA and polypropylene connectors
Interchangeable connectors make an Ace-Thred™ piece of glassware instantly connectable to many tubing Int.Ø's.
Delivery information: Supplied complete with a silicone O-Ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty