Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate, anhydrous 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Carbonic acid potassium saltCarbonic acid, dipotassium salt

AC44762-0010 AC44762-0250 AC44762-0050
TS44762-0050EA 128 USD InStock
TS44762-0050 TS44762-0250 TS44762-0010
Potassium carbonate, anhydrous 98%, Extra Pure
Potassium carbonate


Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg
Order Now
Specification Test Results
Appearance White Crystalline powder
Loss on drying ≤2.0 %
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty