Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC19778-5000 AC19778-0025 AC19778-0100
TS19778-5000EA 158.23 USD InStock
TS19778-5000 TS19778-0025 TS19778-0100
Lithium carbonate 99%, pure
Lithium carbonate


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration excess HCl ≥98.5 %
Loss on ignition ≤0.7 % (500°C, 30 min)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty