Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

AC41326-1000 AC41326-0025
TS41326-0025EA 715 USD InStock
TS41326-0025 TS41326-1000
Lithium carbonate 99+% ACS
Lithium carbonate


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration excess HCl ≥99.0 %
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤5 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤20 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤1000 ppm
Heavy metals (ICP-OES) ≤0.002 %
Sulfur compounds (as SO4) ≤0.2 %
Insoluble matter ≤0.01 % (in dilute HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty