Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubidium chloride 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Synonyms: Rubidiumchloride

AC19392-0100
TS19392-0100EA 285 USD InStock
TS19392-0100
Rubidium chloride 99.99% (trace metals basis)
Rubidium chloride


Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 718 °C
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
Order Now
Specification Test Results
Appearance White Powder and/or chunks
Titration Argentometric 28.9 to 30.1 % (Cl)
Total trace metal impurities ≤100 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty