Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chlorite 80% unstabilized, pure

Supplier: Thermo Scientific Chemicals
Danger

AC22323-0025 AC22323-5000 AC22323-0100 AC22323-0050
TS22323-5000EA 89 USD InStock
TS22323-5000 TS22323-0050 TS22323-0100 TS22323-0025
Sodium chlorite 80% unstabilized, pure
Sodium chlorite


Formula: ClNaO₂
MW: 90.44 g/mol
Melting Pt: 180…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1,II
Merck Index: 12,08743
Order Now
Specification Test Results
Appearance White Powder
Titration Iodometric ≥75 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty