Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

penta-Sodium triphosphate 85% tech., pure

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium triphosphate pentabasicSodium tripolyphosphate

AC21867-5000 AC21867-0025
TS21867-0025EA 149 USD InStock
TS21867-0025 TS21867-5000
penta-Sodium triphosphate 85% tech., pure
Pentasodium triphosphate


Formula: Na₅O₁₀P₃
MW: 367.86 g/mol
Melting Pt: 622 °C
Storage Temperature: Ambient
MDL Number: MFCD00003514
CAS Number: 7758-29-4
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration with HClO4 ≥80 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty