Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfite 98% for biochemistry

Supplier: Thermo Scientific Chemicals

AC44795-2500 AC44795-0010
TS44795-2500EA 44.55 USD InStock
TS44795-2500 TS44795-0010
Sodium sulfite 98% for biochemistry
Sodium sulphite


Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
Order Now
Specification Test Results
Appearance White Powder or crystals
Titration Iodimetric ≥97.5 %
Solubility (1 M in water) Clear colorless
Chloride (Cl) ≤0.02 %
Arsenic (As) ≤1 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Selenium (Se) ≤10 ppm
Zinc (Zn) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty