Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Selenious acid 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Selenous acid

AC43712-0500
TS43712-0500EA 165 USD InStock
TS43712-0500
Selenious acid 99.999% (trace metals basis)
Selenious acid


Formula: H₂O₃Se
MW: 128.97 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
UN: 3283
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White Crystals
Assay 60 to 62.4 % (Se)
Total trace metal impurities ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty