Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenate 98%

Supplier: Thermo Scientific Chemicals
Danger

AC36996-0250 AC36996-5000 AC36996-1000
TS36996-1000EA 231.63 USD InStock
TS36996-1000 TS36996-0250 TS36996-5000
Sodium selenate 98%
Sodium selenate


Formula: Na₂O₄Se
MW: 188.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS Number: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Order Now
Specification Test Results
Appearance White Crystalline powder
Assay ≥97.5 %
Identification confirmed

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty