Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diboron trioxide 98% -40 mesh

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Boron oxideBoric anhydride

AC20630-0250 AC20630-0050 AC20630-0010
TS20630-0050EA 245 USD InStock
TS20630-0050 TS20630-0010 TS20630-0250
Diboron trioxide 98% -40 mesh
di-Boron trioxide


Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C (760 mmHg)
Melting Pt: 450 °C
Density: 2.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 12,01365
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤0.2 %
Titration with NaOH ≥97.5 %
Impurity ≤0.01 % (Total halides impurities)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty