Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilized, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Osmic acid anhydrideOsmic acidOsmium (VIII) oxide

AC42908-1000
TS42908-1000EA 916 USD InStock
TS42908-1000
Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilized, AcroSeal™
Osmium tetraoxide


Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: 40 °C
Density: 0.811 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 1992
ADR: 3,II
Order Now
Specification Test Results
Appearance White to yellow Fused mass or solution
Stabilizer tert-Butyl hydroperoxide
Strength ≥2.5 wt%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty