Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) bromide 98+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Lead dibromide

AC19885-5000 AC19885-1000
TS19885-1000EA 102 USD InStock
TS19885-1000 TS19885-5000
Lead(II) bromide 98+%, Extra Pure
Lead (II) bromide


Formula: Br₂Pb
MW: 367.01 g/mol
Boiling Pt: 916 °C (1013 hPa)
Melting Pt: 373 °C
Density: 6.66 g/cm³
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Titration Argentometric ≥98.0 %
Loss on drying ≤0.1 %
Chloride (Cl) ≤100 ppm
Insoluble matter ≤0.005 % (in acid)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty