Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrichlorosilane 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trichloro(methyl)silane

AC12793-5000 AC12793-0100
TS12793-0100EA 23 USD InStock
TS12793-0100 TS12793-5000
Methyltrichlorosilane 98+%
Methyltrichlorosilane


Formula: CH₃Cl₃Si
MW: 149.48 g/mol
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –77 °C
Density: 1.27 g/cm³
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000481
CAS Number: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,II
REACH: 01-2119475226-36
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty