Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dimethylchlorosilane

AC16284-0500 AC16284-2500 AC16284-0100
TS16284-2500EA 210 USD InStock
TS16284-2500 TS16284-0100 TS16284-0500
Chlorodimethylsilane 97%
Chlorodimethylsilane


Formula: C₂H₇ClSi
Boiling Pt: 34.6 °C (760 mmHg)
Melting Pt: –111 °C
Density: 0.85 g/cm³
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000495
CAS Number: 1066-35-9
EINECS: 213-912-0
UN: 2924
ADR: 3,I
REACH: 01-2119486782-25
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3815 to 1.3855 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty