Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriphenylsilane 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triphenylchlorosilane

AC15125-1000 AC15125-0250
TS15125-0250EA 84 USD InStock
TS15125-0250 TS15125-1000
Chlorotriphenylsilane 95%
Chlorotriphenylsilane


Formula: C₁₈H₁₅ClSi
MW: 294.86 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000496
CAS Number: 76-86-8
EINECS: 200-989-0
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 91 to 96 °C
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty