Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroiodomethane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

AC22215-5000 AC22215-0250 AC22215-1000
TS22215-0250EA 203 USD InStock
TS22215-0250 TS22215-1000 TS22215-5000
Chloroiodomethane 98% stabilized
Chloroiodomethane


Formula: CH₂ClI
MW: 176.38 g/mol
Boiling Pt: 108…109 °C (760 mmHg)
Density: 2.42 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow to pink Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5810 to 1.5840 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty