Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate 99.5%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium metaperiodatePotassium tetroxoiodate (VII)

AC19839-5000 AC19839-0025 AC19839-1000
TS19839-5000EA 335 USD InStock
TS19839-5000 TS19839-1000 TS19839-0025
Potassium periodate 99.5%, Extra Pure
Potassium periodate


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 582 °C
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07823
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Titration Iodometric ≥99.4 %
Other halogens (as Cl) ≤0.01 %
Manganese (Mn) ≤1 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty