Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium metaperiodatePotassium tetroxoiodate (VII)

AC41829-5000 AC41829-1000
TS41829-5000EA 344 USD InStock
TS41829-5000 TS41829-1000
Potassium periodate ACS
Potassium periodate


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 582 °C
Storage Temperature: Ambient
MDL Number: MFCD00011407
CAS Number: 7790-21-8
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration with Na2S2O3 99.8 to 100.3 % (dried basis)
Other halogens (as Cl) ≤0.01 %
Manganese (Mn) ≤1 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty