Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdic acid ACS

Supplier: Thermo Scientific Chemicals
Warning

AC42398-1000 AC42398-5000
TS42398-5000EA 210.63 USD InStock
TS42398-5000 TS42398-1000
Molybdic acid ACS
Molybdic acid


Formula: H₂MoO₄
MW: 161.95 g/mol
Density: 3.1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 12,06322
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Titration ≥85.0 % (Precipitation titraton of molybdate)
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.2 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.01 % (in dilute NH4OH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty