Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth(III) nitrate pentahydrate 98% for analysis

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bismuth trinitrate pentahydrate

AC40261-5000 AC40261-0025
TS40261-0025EA 397 USD InStock
TS40261-0025 TS40261-5000
Bismuth(III) nitrate pentahydrate 98% for analysis
Bismuth (III) nitrate pentahydrate


Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Arsenic (As) ≤10 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤20 ppm
Silver (Ag) ≤10 ppm
Insoluble matter ≤0.005 % (in HNO3)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty