Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium oxide 90%, powder, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC20783-0250 AC20783-0025 AC20783-5000
TS20783-0250EA 29 USD InStock
TS20783-0250 TS20783-5000 TS20783-0025
Barium oxide 90%, powder, Extra Pure
Barium oxide


Formula: BaO
MW: 153.33 g/mol
Boiling Pt: 2000 °C (760 mmHg)
Melting Pt: 1920 °C
Storage Temperature: Ambient
MDL Number: MFCD00003453
CAS Number: 1304-28-5
EINECS: 215-127-9
UN: 1884
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to beige to yellow-green to gray Powder
Titration with HCl ≥88 %
Barium carbonate ≤11 %
Insoluble matter ≤0.5 % (in HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty