Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony (III) sulfide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diantimony trisulfideAntimony trisulfideDiantimony trisulphideAntimony trisulphideStibnite

AC22359-5000 AC22359-1000
TS22359-5000EA 121 USD InStock
TS22359-5000 TS22359-1000
Antimony (III) sulfide 98%
Antimony (III) sulphide


Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150 °C (1013 hPa)
Melting Pt: 550 °C
Storage Temperature: Ambient
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
Order Now
Specification Test Results
Appearance Dark gray to black Crystalline powder
Assay ≥97.5 % (Sb2S3)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty