Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(II) sulfide, fused sticks

Supplier: Thermo Scientific Chemicals
Synonyms: Ferrous sulphideFerrous sulfide

AC23330-0010
TS23330-0010EA 61 USD InStock
TS23330-0010
Iron(II) sulfide, fused sticks
Iron (II) sulphide


Formula: FeS
MW: 87.91 g/mol
Melting Pt: 1195 °C
Density: 4.84 g/cm³
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
Order Now
Specification Test Results
Appearance Gray-black Sticks
Assay (S) about 25%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty