Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cesium sulfate 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC21870-0250 AC21870-0100 AC21870-2500 AC21870-1000
TS21870-0250EA 64 USD InStock
TS21870-0250 TS21870-0100 TS21870-2500 TS21870-1000
Cesium sulfate 99+%, pure
Cesium sulphate


Formula: Cs₂O₄S
MW: 361.87 g/mol
Boiling Pt: 1900 °C (760 mmHg)
Melting Pt: 1005 °C
Storage Temperature: Ambient
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Assay ≥99.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty