Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Diphenyl-1,1,2,2-tetramethyldisilane 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,2-Diphenyltetramethyldisilane1,1,2,2-Tetramethyl-1,2-diphenyldisilane

AC38788-0010
TS38788-0010EA 57 USD InStock
TS38788-0010
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 97%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane


Formula: C₁₆H₂₂Si₂
MW: 270.52 g/mol
Boiling Pt: 73 °C (0.3 mmHg)
Melting Pt: 33…38 °C
Flash Pt: 105 °C
MDL Number: MFCD00054786
CAS Number: 1145-98-8
Order Now
Specification Test Results
Appearance White to light brown Low melting mass
Infrared spectrum Conforms
Melting point 33 to 38 °C
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty