Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) acetate tetrahydrate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid manganese (II) salt tetrahydrateManganese acetate tetrahydrateManganese diacetate tetrahydrate

AC21256-0500
TS21256-0500EA 149 USD InStock
TS21256-0500
Manganese(II) acetate tetrahydrate 99.999% (trace metals basis)
Manganese (II) acetate tetrahydrate


Formula: (H₃CCOO)₂Mn·4H₂O
MW: 245.09 g/mol
Melting Pt: >300 °C
Flash Pt: >130 °C
Storage Temperature: Ambient
MDL Number: MFCD00062552
CAS Number: 6156-78-1
Order Now
Specification Test Results
Appearance Pink Crystalline powder or crystals
Titration Complexometric 21.8 to 23.0 %
Total trace metal impurities ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty