Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium oxalate 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium oxalateSodium ethanedioateSoerensen's buffer substances

AC27054-0010
TS27054-0010EA 167 USD InStock
TS27054-0010
Disodium oxalate 99.5% for analysis
di-Sodium oxalate


Formula: Na₂C₂O₄
MW: 134 g/mol
Melting Pt: 250…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 12,08795
Order Now
Specification Test Results
Appearance White Powder
Loss on drying ≤0.05 % (105°C)
pH 7 to 8.5 (2.5 % at 20°C)
Titration with KMnO4 ≥99.5 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Ammonium (NH4) ≤20 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty