Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitroindole 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Nitro-1H-indole

AC12870-0250 AC12870-0050
TS12870-0250EA 331 USD InStock
TS12870-0250 TS12870-0050
5-Nitroindole 99%
5-Nitroindole


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD00005673
CAS Number: 6146-52-7
EINECS: 228-153-0
Order Now
Specification Test Results
Appearance Yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 140 to 143 °C
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty