Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(I) oxide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cuprous oxideDicopper oxide

AC19742-0250 AC19742-0025 AC19742-5000
TS19742-0025EA 239 USD InStock
TS19742-0025 TS19742-0250 TS19742-5000
Copper(I) oxide 97%
Copper (I) oxide


Formula: Cu₂O
MW: 143.09 g/mol
Boiling Pt: 1800 °C (1013 hPa)
Melting Pt: 1232 °C
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Red-brown Powder
Titration Complexometric ≥96.0 %
Iron (Fe) ≤500 ppm
Nickel (Ni) ≤100 ppm
Silver (Ag) ≤50 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty