Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethyldisilane 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC20269-2500 AC20269-0100 AC20269-0500
TS20269-2500EA 585 USD InStock
TS20269-2500 TS20269-0500 TS20269-0100
Hexamethyldisilane 98+%
Hexamethyldisilane


Formula: C₆H₁₈Si₂
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.71 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4211 to 1.4231 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty